Learn More
Velp Scientifica™ Imhoff Cone
SDP
Catalog No. 11392158
Change view
Click to view available options
Material:
Glass
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-392-158 Plastic
11-392-159 Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-392-158 Supplier Velp Scientifica™ Supplier No. A00001002
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Velp Scientifica™ Imhoff Cone for use with FCS and JLT Series Flocculators.
  • Transparent for visability
  • Available in glass or plastic
  • Available in graduated
  • For use with Flocculators
Specifications
Capacity (Metric) 1000 mL
For Use With (Equipment) FCS, JLT
Type Imhoff Cone
Material Plastic