Learn More
Apexbio Technology LLC Higenamine hydrochloride 11041-94-4 100mg
SDP
Supplier:  Apexbio Technology LLC C6665100
Encompass_Preferred
Higenamine hydrochloride (CAS 11041-94-4) is a plant-derived alkaloid primarily isolated from aconite species This compound demonstrates selective inhibition of histone demethylase LSD1 thereby influencing epigenetic regulation processes Higenamine hydrochloride has been shown to modulate reactive oxygen species (ROS)-related PI3K/Akt signaling pathways and suppress IL-1 -induced apoptosis as well as to activate SMAD2/3 signaling to facilitate osteogenic differentiation It is utilized in biomedical research to investigate mechanisms underlying tumor progression inflammatory responses hypoxic neural injury and cardiorenal syndrome
Catalog No. 50-000-92731
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.