Learn More
Cayman Chemical ImIdafenacIn 25mg
SDP
Supplier:  Cayman Chemical 3742025 MG
Encompass_Preferred
A dual inhibitor of COX and 5-LO (IC50s 2.84 and 0.15 UM respectively in RBL-1 cell lysates) inhibits the activity of 12- but not 15- LO (IC50s 4.45 and 146.6 UM for the human platelet and rabbit reticulocyte enzymes respectively) reduces A23187-induced TXB2 and LTB4 production in human PBMCs (IC50s 0.01 and 0.07 UM respectively) and decreases epinephrine-induced platelet aggregation in human PRP (IC50 0.045 UM) inhibits paw swelling in a rat model of adjuvant-induced arthritis (ED50 3.5 mg/kg)
Catalog No. 50-291-8165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.