Learn More
Sigma Organic Chemistry Imidazo 1 2-apyridin-2- | 1G | 82090-52-6 | MFCD06797471
Supplier: Sigma Organic Chemistry SYX003801G

Imidazo 1 2-apyridin-2- , 1G
About this item:
CAS #: 82090-52-6
MDL #: MFCD06797471
Chemical Formula: C8H8N2O
Molecular Weight: 148.16
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.