Learn More
Selleck Chemical LLC Imidazole ketone erastin2mg
SDP
Supplier:  Selleck Chemical LLC S88772MG
Encompass_Preferred
Imidazole ketone erastin (PUN30119, IKE) is a potent, selective, and metabolically stable system xc¿ inhibitor and inducer of ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3414
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.