Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazolidinyl Urea, MP Biomedicals™

Catalog No. ICN15500980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15500980 100 g
ICN15500925 25 g
ICN15500983 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15500980 Supplier MP Biomedicals, Inc Supplier No. 0215500980
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39236-46-9
Molecular Formula C11H16N8O8
Molecular Weight (g/mol) 388.297
MDL Number MFCD00221482
InChI Key ZCTXEAQXZGPWFG-UHFFFAOYSA-N
Synonym imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
PubChem CID 38258
ChEBI CHEBI:51805
IUPAC Name 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
SMILES C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO

Specifications

Color White
Quantity 100 g
Formula Weight 388.29
Physical Form Powder
Chemical Name or Material Imidazolidinyl urea, Preservative, antioxidants
EINECSNumber 254-372-6
Recommended Storage Store at +4°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.