Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminodiacetic Acid, Disodium Salt Monohydrate 97%, Thermo Scientific™

Catalog No. AC122061000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC122061000 100 g
AC122065000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122061000 Supplier Thermo Scientific Chemicals Supplier No. 122061000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17593-73-6
Molecular Formula C4H5NNa2O4
Molecular Weight (g/mol) 177.07
MDL Number MFCD00150689
InChI Key HAXVIVNBOQIMTE-UHFFFAOYSA-L
Synonym disodium iminodiacetate hydrate, iminodiacetic acid disodium salt hydrate, sodium 2,2'-azanediyldiacetate hydrate, disodium hydrate iminodiacetate, iminodiacetic acid, disodium salt monohydrate, sodium iminodiacetate dibasic hydrate, acmc-209ri5, c4h5no4.2na.h2o, disodium iminodiacetate monohydrate, sodium 2,2-azanediyldiacetate hydrate
PubChem CID 12520792
IUPAC Name disodium 2-[(carboxylatomethyl)amino]acetate
SMILES [Na+].[Na+].[O-]C(=O)CNCC([O-])=O

Specifications

Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula HN(CH2CO2Na)2·H2O
Quantity 100 g
Beilstein 04, 365
Merck Index 10, 4815
Formula Weight 195.08
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material Iminodiacetic acid, disodium salt monohydrate, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.