Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminodibenzyl, 97%

Catalog No. AAA1920436
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1920436 500 g
AAA1920414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1920436 Supplier Thermo Scientific Chemicals Supplier No. A1920436

CAS: 494-19-9 | C14H13N | 195.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-19-9
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00005070
InChI Key ZSMRRZONCYIFNB-UHFFFAOYSA-N
Synonym iminodibenzyl, 10,11-dihydro-5h-dibenzo b,f azepine, 10,11-dihydro-5h-dibenz b,f azepine, iminobibenzyl, 2,2'-iminodibenzyl, 2,2'-iminobibenzyl, iminodibenyl, 5h-dibenz b,f azepine, 10,11-dihydro, unii-262bx7oe3u, 10,11-dihydro-5-dibenz b,f azepine
PubChem CID 10308
SMILES C1CC2=CC=CC=C2NC2=CC=CC=C12

Specifications

Melting Point 105°C to 108°C
Odor Odorless
Quantity 500 g
Beilstein 152732
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Formula Weight 195.27
Percent Purity 97%
Chemical Name or Material Iminodibenzyl
RTECSNumber HN8950000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.