Learn More
U.S. Pharmacopeia Imipenem monohydrate, 74431-23-5, MFCD09753321, 100 mg
Supplier:  U.S. Pharmacopeia 1337809
Encompass
Empirical Formula (Hill Notation): C12H17N3O4S · H2O, Molecular Weight: 317.36, Synonym: (5R,6S)-6-[(1R)-1-Hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid monohydrate, Primaxin monohydrate, Tienam monohydrate.
Catalog No. NC9022260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.