Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Imipramine Monoclonal Antibody (IM11-214.1)
GREENER_CHOICE

Catalog No. 01671952 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-952 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-952 Supplier Invitrogen™ Supplier No. MA110722
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-10722 detects Imipramine. MA1-10722 has been successfully used in ELISA applications. The MA1-10722 immunogen is Imipramine, conjugated to KLH.

Imipramine Base is a synthetic tricyclic derivative, antidepressant Imipramine enhances monoamine neurotransmission in certain areas of the brain. It also induces sedation through histamine 1 receptor blockage; hypotension through beta-adrenergic blockage; and diverse parasympatholytic effects. Imipramine has less sedative effect than other members of its therapeutic family. It is used in major depression, dysthymia, bipolar depression, attention-deficit disorders, agoraphobia, and panic disorders. [National Center for Biotechnology Information. PubChem Compound Database; CID=3696].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Imipramine
Applications ELISA
Classification Monoclonal
Clone IM11-214.1
Concentration ~1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Antideprin; C19H24N2; Deprenil; Deprimin; Deprinol; Depsonil; Dynaprin; Eupramin; Imipramil; Irmin; Janimine; Melipramin; Surplix; Tofranil
Host Species Mouse
Immunogen Imipramine, conjugated to KLH.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.