Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Imiquimod, TLR7 Ligand
SDP

Catalog No. NBP2262281 Shop All Novus Biologicals Products
Change view
Click to view available options
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP2262281 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2262281 Supplier Novus Biologicals™ Supplier No. NBP2262281MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Applications: Functional, In vitro assay, Ligand Activation

Imiquimod, TLR7 ligand is an imidazoquinoline amine analog to guanosine. As a synthetic molecule of the Imidazoquinoline family, it has potent immunostimulatory activity. Imiquimod has been shown to activate only TLR7, this activation is MyD88 dependant and leads to the induction of the transcription factor NF-kB.

  • Species: Human, Mouse
  • Specificity: Imiquimod is human/mouse TLR7 agonist
  • Stimulation of TLR7 can be achieved with 5-10μg/mL

Recommended Storage

Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.

Specifications

Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
For Use With (Application) For activation of TLR7
Product Type Imiquimod, TLR7 Ligand
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.