Learn More
Description
Specifications
Specifications
| Concentration | 2.5 mg/mL |
| Formulation | C14H16N4, HCl |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Quantity | 0.125 mg |
| For Use With (Application) | This product is useful for activation of TLR7 and stimulation of TLR7 has been achieved with 5 to 10μL. Use in ligand activation, functional, and in vitro reported in scientific literature (PMID 25957979) |
| Target | Imiquimod, TLR7 ligand |
| Species | Human/Mouse |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
