Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Imiquimod, Toll-like Receptor 7 Agonist, Solid, 100 mg

Catalog No. 502706639
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6639 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6639 Supplier Abcam Supplier No. AB141410100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent Toll-like receptor 7 agonist with immunomodulatory, antiviral, and antitumor activity.

  • Molecular weight: 240.3 Da
  • Purity: >95%
  • Form: Solid
  • Mechanism: Toll-like receptor 7 agonist; activates NF-κB and stimulates pro-inflammatory cytokines
  • Applications: Immunomodulator, antiviral and antitumor agent; active in vivo and in vitro
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice

Specifications

Target Molecule Toll-like Receptor 7
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Imiquimod
Quantity 100 mg
For Use With (Application) Immunomodulation, antiviral research, antitumor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.