Learn More
Abcam Imiquimod, Toll-like Receptor 7 Agonist, Solid, 500 mg
Description
- Molecular weight: 240.3 Da
- Purity: >95%
- Form: Solid
- Activity: Toll-like receptor 7 agonist; activates NF-κB and stimulates pro-inflammatory cytokines
- Applications: Immunomodulator, antiviral and antitumor agent
- Usage: Active in vivo and in vitro
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Toll-like Receptor 7 |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 500 mg |
| For Use With (Application) | Immunomodulation, antiviral research, antitumor research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.