Learn More
Description
- The ImmEdge™ Hydrophobic Barrier PAP Pen creates a water-repellent, heat-stable barrier to keep reagents localized on tissue specimens, which prevents mixing when multiple sections are mounted on a single slide.
- Applications: Functional assay, immunocytochemistry/immunofluorescence, immunohistochemistry, and in-situ hybridization
- Storage: Store at room temperature
Specifications
Specifications
| Product Type | PAP Pen |
| Quantity | 2 Pens |
| For Use With (Application) | Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence, ISH |
| Product Line | Novus |
| Content And Storage | Store at room temperature. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
