Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Immethridine dihydrobromide, Tocris Bioscience™
SDP

Catalog No. 231550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-155-0 50 mg
23-151-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-155-0 Supplier R&D Systems Supplier No. 2315/50
This item is not returnable. View return policy

Potent H3 agonist, highly selective over H4

Potent and highly selective histamine H3 receptor agonist (pEC50 = 9.74) that displays 300-fold selectivity over the H4 receptor (pKi values are 9.07 and 6.61 respectively). Does not bind to H1 or H2 receptors at concentrations up to 10 μM. .

Chemical Identifiers

CAS 699020-93-4
Molecular Formula C9H11Br2N3
Molecular Weight (g/mol) 321.016
InChI Key CYNKWHIKNDIVDR-UHFFFAOYSA-N
Synonym immethridine dihydrobromide, 4-1h-imidazol-4-ylmethyl pyridine dihydrobromide, 4-1h-imidazol-5-ylmethyl pyridine;dihydrobromide, 4-1h-imidazol-5-yl methyl pyridine-hydrogen bromide 1/2
PubChem CID 16759172
IUPAC Name 4-(1H-imidazol-5-ylmethyl)pyridine;dihydrobromide
SMILES C1=CN=CC=C1CC2=CN=CN2.Br.Br

Specifications

Quantity 50 mg
Formula Weight 321.01
Percent Purity >99%
Chemical Name or Material Immethridine dihydrobromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.