Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Immobilon ECL Ultra Western HRP Substrate

Catalog No. p-7138965 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WBULS0100 100 mL
WBULS0500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. WBULS0100 Supplier MilliporeSigma™ Supplier No. WBULS0100
Only null left
Add to Cart
Add to Cart

Includes

Immobilon ECL Ultra Western HRP Substrate 100ML: Solution A- 50 mL; Solution B- 50 mL

MilliporeSigma™ Immobilon ECL Ultra Western HRP Substrate excels in all Western and dot/slot/spot blotting applications. Compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.

Features:

  • Immobilon™ ECL Ultra Western HRP Substrate is an ultra-sensitive reagent with long signal duration for the detection of horseradish peroxidase (HRP)-labeled antibodies on western blots.
  • The intense signal created permits sensitive detection at the mid-to-low femtogram range using x-ray film or digital imaging systems.
  • Immobilon™ ECL Ultra substrate is compatible with both PVDF and nitrocellulose blotting membranes, as well as commonly used buffers and blocking reagents.
  • Sufficient for 1000 cm²

Certifications

request

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western blotting, HRP
Quantity 100 mL

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.