Learn More
Aviva Systems Biology Corp IMMP2L Antibody - C-terminal region (ARP60563_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60563_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IMMP2L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human IMMP2L. Alternate names: IMP2, IMP2-LIKEApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human
Catalog No. 50-287-085