Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ImmPACT (TM) DAB EqV HRP Substrate
SDP

Catalog No. NB119927 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119927 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119927 Supplier Novus Biologicals™ Supplier No. SK4103NB100ML
Add to Cart
Add to Cart

Optimized for IHC/ICC/ISH

Specifications

Quantity 100 mL
Substrate Type DAB Substrate
Item Description ImmPACT(TM) DAB EqV (equal volume) consists of 200 ml each of Reagent 1 and Reagent 2 which when mixed together in equal volumes form the working solution. ImmPACT(TM) DAB EqV produces a dark brown reaction product that is 3 to 5 times more sensitive than the original DAB substrate. The working solution is stable for several days when stored at 4°C. This substrate can be used for both manual and automated staining methods. ImmPACT(TM) DAB EqV can be used for blot detection on nylon and PVDF membranes but is not recommended for nitrocellulose membranes. Sections stained with DAB can also be viewed by darkfield and electron microscopy. Slides developed with DAB can be dehydrated, cleared, and permanently mounted.
Storage Requirements Store at 4°C in the dark.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.