Learn More
Vector Laboratories Streptavidin, DyLight 488, 1mg MP-7602-50
SDP
Supplier:  Vector Laboratories MP760250
Encompass_Preferred
Streptavidin, DyLight 488, 1 mg. DyLight 488 Streptavidin can be used to detect biotinylated secondary antibodies and other macromolecules in applications such as immunofluorescence, in situ hybridization, or flow cytometry. Advantages of DyLight dyes include greater photostability, pH independence, and brighter fluorescence. Excitation maximum: 493 nm; Emission maximum: 518 nm; Color: Green.
Catalog No. 50-204-5689
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.