Learn More
Vector Laboratories ImmPACT SG HRP Substrate 120ml MP-7601-50
SDP
Supplier:  Vector Laboratories MP760150
Encompass_Preferred
ImmPACT SG Substrate, Peroxidase (HRP), 120 ml. The ImmPACT SG HRP substrate produces a blue-gray reaction product that can be used as a single label or as a second color for multiple antigen labeling. It is 2–4 times more sensitive than original substrate kit. Features: Optimized for IHC/ICC/ISH and blots, Permanent mounting, Working solutions stable for up to 14 days at 4 degrees C, Supplied as 120 ml of diluent and stock solutions in dropper bottles.
Catalog No. NC1692272
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.