Learn More
Novus Biologicals™ ImmPRESS(TM) HRP Horse anti-Mouse IgG PLUS Polymer, Peroxidase Kit

Specifications
Specifications
| Target Molecule | ImmPRESS(TM) HRP Horse anti-Mouse IgG PLUS Polymer, Peroxidase |
| Quantity | 1 Kit |
| Content And Storage | Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.