Learn More
R&D Systems™ ImmPRESS(TM) HRP Universal (Horse anti-Mouse/Rabbit IgG) PLUS Polymer, Peroxidase Kit
Shop All Novus Products

Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Product Line | Novus |
| Target Molecule | ImmPRESS(TM) HRP Universal (Horse anti-Mouse/Rabbit IgG) PLUS Polymer, Peroxidase |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.