Learn More
Amsbio LLC Immune multiple organs tumor and normal tissue array
Supplier:  Amsbio LLC MC1081
Encompass
Immune multiple organs tumor and normal tissue array containing 20 cases leukemia 20 cases non-Hodgkin's lymphoma 20 cases Hodgkin's lymphoma 11 cases plasma cell myeloma 9 cases extramedullary plasmacytoma 20 cases malignant thymoma plus 2 each cases of normal lymph node spleen bone marrow and thymus gland tissue 108 cases/108 cores
Catalog No. 50-256-4167