Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Clear Standard Modules
GSA_VAGREENER_CHOICE

Catalog No. 1424581
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
PolySorp
Volume (Metric) Working:
250 μL
330 μL
350 μL
Well Shape:
Round
Round, C Bottom
Round, Flat Bottom (F-well)
Well Design:
C12
C8
F12
F16 (2x8 wells)
F8
U8
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Volume (Metric) Working Well Shape Well Design
14-245-81 Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
22-047029 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-567-130 MaxiSorp 250 μL Round, C Bottom C8
12-565-265 MaxiSorp 250 μL Round, C Bottom C8
12-565-119 MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
12-565-121 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-565-122 PolySorp 350 μL Round, Flat Bottom (F-well) F8
12-565-127 MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
12-565-129 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-565-130 PolySorp 350 μL Round, Flat Bottom (F-well) F8
12-565-123 MaxiSorp 250 μL Round, C Bottom C12
12-565-117 MaxiSorp 250 μL Round U8
12-565-118 PolySorp 250 μL Round U8
14-245-167 Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
14-245-193B Immulon 4 HBX 330 μL Round, Flat Bottom (F-well) F12
21-377-254 Enhanced Binding 350 μL Round, Flat Bottom (F-well) F8
14-245-80 Immulon 1 B 330 μL Round, Flat Bottom (F-well) F12
12-565-124 PolySorp 250 μL Round, C Bottom C12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 14-245-81 Supplier Thermo Scientific™ Supplier No. 6302
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat-bottom, C-bottom (radius edge), U-shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

Certifications

Products specified in chart are certified for: Adsorption of IgG; CV of <5% between wells; all results +/-10% from mean; and background of all wells +/-0.005 absorbance units from mean.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart Breakable Strips
Binding Property Slightly Hydrophilic
Certifications/Compliance Certified for adsorption of IgG; CV of <5% between wells
Certified Yes
Format Strip Plate
Lid With Lid
Detection Method Colorimetric
Packaging 320 Pk., 320 Cases
Binding Type Adsorption
Color Clear
Product Type Microplate Strip
Material Polystyrene
Volume (Metric) Well 380 μL
No. of Wells 96
Volume (Metric) Working 330 μL
Surface Treatment Immulon 2 HB
Well Shape Round, Flat Bottom (F-well)
No. per Case 320
No. per Pack 320
Plate Blocking None
Quantity 320/Pk.
Target Molecule Proteins
Well Design F12
Show More Show Less
What frames do you recommend for your Immuno Clear Standard Modules?

We recommend Frames and Accessories for Immuno Breakable Modules, BreakApart frame (Cat. No. 465404). This frame is designed to hold the Immuno Clear Standard Modules.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Which Nunc immunodiagnostic passive surface should I choose: PolySorp, MediSorp, MaxiSorp, or MultiSorp?

Our passive immunodiagnostic surfaces are based on a series of treatments that modify the polystyrene surface to different final levels of hydrophilicity. Selection of one of these surfaces for ELISA development should be based on the characteristics of the biomolecule you seek to immobilize. PolySorp is fairly hydrophobic and should be used for biomolecules that have hydrophobic domains, e.g., lipids, lipoproteins, and large proteins. MediSorp is slightly hydrophilic and can be used for biomolecules with hydrophilic and hydrophobic properties, e.g., medium-to-large proteins such as albumin, or amphiphilic biomolecules such as LPS. MaxiSorp is quite hydrophilic, optimized for binding of IgG (and any other molecule with similar characteristics) and thus is a popular choice for sandwich ELISA and similar protocols. MultiSorp is very hydrophilic, and in turn will immobilize very hydrophilic biomolecules such as glycoproteins. Some optimization will always be necessary, and in many cases screening for binding optimization may involve a few surface types.

What is the difference between a certified and an uncertified immunodiagnostic plate, e.g., Cat. No. 439454 vs. 442404?

There is no difference in the manufacturing process. Certified plates are performance tested (using an antibody binding protocol) during quality control, whereas uncertified plates are not. This test is expensive for us to perform, so we offer the uncertified version at cost savings customers for whom antibody binding testing results are not critical.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.