Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Catalog No. 12565193
Change view
Click to view available options
Material:
Polyethylene
Polystyrene
Diameter (Metric) Outer:
12 mm
15 mm
Product Type:
Immuno Tubes and Stopper
Stopper
For Use With (Equipment):
12 mm Nunc-Immuno Tubes
15 mm Nunc-Immuno Tubes
Immuno Tube
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Diameter (Metric) Outer Product Type For Use With (Equipment)
12-565-193 Polyethylene 15 mm Stopper 15 mm Nunc-Immuno Tubes
12-565-195 Polyethylene 12 mm Stopper 12 mm Nunc-Immuno Tubes
12-565-148 Polyethylene 12 mm Immuno Tubes and Stopper Immuno Tube
12-565-146 Polystyrene 12 mm Immuno Tubes and Stopper Immuno Tube
12-565-150 Polyethylene 15 mm Immuno Tubes and Stopper Immuno Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Catalog No. 12-565-193 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 343036
Add to Cart
Add to Cart

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.

Features:

  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment

Choice of Surface:

  • Thermo Scientific™ PolySorp™: a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains
  • Thermo Scientific™ MaxiSorp™: a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character
  • Thermo Scientific™ Mini-Sorp™: a polyethylene-based material that provides a low-binding surface

Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):

  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents or collecting chromatographic fractions

Solid Phase Immunoassay Tubes:

  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • Certified binding homogeneity
  • High-quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times

Recommended for:

  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stopper
Material Polyethylene
Diameter (Metric) Outer 15 mm
No. per Pack 300
No. per Case 2400
Volume (Metric) Not Applicable
For Use With (Equipment) 15 mm Nunc-Immuno Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.