Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Immunoblot Blocking Reagent

Catalog No. 20200MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20200MI 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20200MI Supplier MilliporeSigma™ Supplier No. 20200

missing translation for 'validMainframeMsg'

For use in western blots as a blocking agent

  • Non-fat dry milk fortified with vitamin A palmitate and vitamin D2
  • Contains no preservatives
  • Quantity: 20g

Recommended Storage

Store at 4°C.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Immunoblot Blocking Reagent
For Use With (Application) Western Blot
Quantity 20 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.