Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Grass Pollen Allergens

Catalog No. 14410001P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
g1
g2
g3
g4
g5
g6
g7
g8
g9
g10
g11
g12
g13
g14
g15
g16
g17
g70
g71
g201
g202
Common Name:
Bahia grass
Barley
Bermuda grass
Brome grass
Canary grass
Cocksfoot
Common reed
Cultivated oat
Cultivated rye
Cultivated wheat
Johnson grass
Maize, Corn
Meadow fescue
Meadow foxtail
Meadow grass, Kentucky blue
Redtop, Bentgrass
Rye-grass
Sweet vernal grass
Timothy grass
Velvet grass
Wild rye grass
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name
14-410-001P 16 Tests g6 Timothy grass
14-413-401P 16 Tests g5 Rye-grass
14-413-601P 16 Tests g8 Meadow grass, Kentucky blue
14-410-101P 16 Tests g12 Cultivated rye
14-411-701P 16 Tests g3 Cocksfoot
14-413-001P 16 Tests g1 Sweet vernal grass
14-413-301P 16 Tests g4 Meadow fescue
14-413-101P 16 Tests g2 Bermuda grass
14-413-901P 16 Tests g13 Velvet grass
14-413-801P 16 Tests g10 Johnson grass
14-413-701P 16 Tests g9 Redtop, Bentgrass
14-418-801P 16 Tests g15 Cultivated wheat
14-418-701P 16 Tests g7 Common reed
14-418-901P 16 Tests g17 Bahia grass
14-423-801P 16 Tests g11 Brome grass
14-424-001P 16 Tests g16 Meadow foxtail
14-423-901P 16 Tests g14 Cultivated oat
14-434-401P 10 Tests g70 Wild rye grass
14-436-001P 10 Tests g71 Canary grass
14-482-101P 10 Tests g201 Barley
14-482-201P 10 Tests g202 Maize, Corn
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
This item is not returnable. View return policy
Catalog No. 14-410-001P Supplier Thermo Scientific™ Supplier No. 14410001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with ImmunoCAP™ Grass Pollen Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option
Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms
As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Scientific Name Phleum pratense
Regulatory Status FDA Cleared
Code Type Grass
Common Name Timothy grass
Quantity 16 Tests
Code g6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.