Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Spice Allergens

Catalog No. 14504410P
Change view
Click to view available options
Code:
Rf220
Rf234
Rf263
Rf265
Rf266
Rf268
Rf269
Rf270
Rf271
Rf272
Rf273
Rf274
Rf277
Rf278
Rf279
Rf282
Rf283
Rf317
Rf332
f218
f280
f86
f89
Common Name:
Anise
Basil
Bay leaf
Black pepper
Caraway
Chilipepper
Cinnamon
Clove
Coriander
Dill
Ginger
Green pepper (unripe seed)
Mace
Marjoram
Mint
Mustard
Nutmeg
Oregano
Paprika, Sweet pepper
Parsley
Tarragon
Thyme
Vanilla
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Code Common Name
14-504-410P Rf271 Anise
14-429-701P f86 Parsley
14-432-901P f89 Mustard
14-481-601P f218 Paprika, Sweet pepper
14-481-701P f280 Black pepper
14-504-910P Rf278 Bay leaf
14-504-810P Rf269 Basil
14-505-710P Rf265 Caraway
14-506-310P Rf220 Cinnamon
14-506-410P Rf268 Clove
14-506-910P Rf277 Dill
14-507-210P Rf270 Ginger
14-507-510P Rf263 Green pepper (unripe seed)
14-508-310P Rf266 Mace
14-508-410P Rf274 Marjoram
14-508-710P Rf282 Nutmeg
14-508-810P Rf283 Oregano
14-510-310P Rf272 Tarragon
14-510-610P Rf234 Vanilla
14-510-510P Rf273 Thyme
14-521-410P Rf317 Coriander
14-524-910P Rf332 Mint
14-506-210P Rf279 Chilipepper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
This item is not returnable. View return policy
Catalog No. 14-504-410P Supplier Thermo Scientific™ Supplier No. 14504410
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with the ImmunoCAP™ Spice Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Scientific Name Pimpinella anisum
Regulatory Status ASR
Code Type Spice
Common Name Anise
Quantity 10 Tests
Code Rf271
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.