Learn More
Drg International Inc Immunoglobulin E (Ige) Elisa
Supplier:  Drg International Inc EIA1788
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of immunoglobulin E (IgE) concentration in serum. Patients with atopic allergic diseases such as atopic asthma, atopic dermatitis, and hay fever have been shown to exhibit increased total Immunoglobulin E (IgE) levels in blood. The DRG IgE Quantitative Test is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The test specimen (serum) is added to the IgE monoclonal antibodies immobilized on polystyrene microtiter wells (solid phase) and incubated with the Zero Buffer. If human IgE is present in the specimen, it will combine with the antibodies on the well.
Catalog No. 50-194-7541