Learn More
Electron Microscopy Sciences Aurion ImmunoGold Ultra Small Gar/Gam/BSA-C/R-Gent Kit
SDP
Supplier:  Electron Microscopy Sciences 25550
Encompass_Preferred
Immunogold reagent kit. Affinity purified and cross adsorbed antibodies. Includes: 0.6ml Goat-anti-Rabbit GP-Ultra Small 0.6ml, Goat-anti-Mouse IgG/IgM GP-Ultra-Small, 30ml BSA-c background agent, R-Gent Silver Enhancement.
Catalog No. 50-282-91