Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMMY CrAg Lateral Flow Assay
SDP

Catalog No. 22122320
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Cryptococcus Antigen Assay Strips
Cryptococcus Antigen LF Specimen Diluent
Quantity:
50 Tests
6.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
22-122-320 Cryptococcus Antigen Assay Strips 50 Tests
22-122-321 Cryptococcus Antigen LF Specimen Diluent 6.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Always wear gloves when handling reagents.
Catalog No. 22-122-320 Supplier IMMY Supplier No. CR2003US
Add to Cart
Add to Cart
This item is not returnable. View return policy
Always wear gloves when handling reagents.

Requires

Pipetter (40 and 80μL); timer; disposable micro-centrifuge tubes, test tubes or microtiter plate

Prescription-use laboratory assay aids in the diagnosis of crytococcosis

Immochromatographic test system for qualitative or semi-quantitative detection of capsular antigens of Cryptococcus species complex in serum and cerebral spinal fluid.

  • No specimen treatment needed
  • 10-Minute total test time
  • High sensitivity and specificity — twice as fast, and with greater sensitivity, compared to latex agglutination
Dipstick sandwich immunochromatographic assay
  • Specimens and specimen diluent are added into a test tube or other reservoir
  • Lateral flow device is placed into the reservoir
  • Specimen wicking captures gold-conjugated, anti-CrAg monoclonal antibodies and gold-conjugated control antibodies deposited on test membrane
  • CrAg present in the specimen binds to the gold-conjugated, anti-CrAg antibodies
  • Antibody-antigen complex continues to wick up the membrane and interacts with test line containg immobilized anti-CrAg monoclonal antibodies
  • Gold-labeled antibody-antigen complex forms a sandwich at the test line causing a visible line to form
  • Positive test results create two lines (test and control)
  • Negative test results form only one line (control)
  • Failed test exists if a control line fails to develop

Assay Kit Content

  • 6.5mL LF Specimen Diluent: Glycine-buffered saline containing blocking agents and a preservative
  • Pack of 50 CrAg LF Test Strips in desiccant vial
  • 1mL CrAg Positive Control: Glycine- buffered saline spiked with cryptococcal antigen (strain 184A – clinical isolate from Tulane University (Infection & Immunity, June 1983, p. 1052-1059))
  • Package insert

For optimal results, sterile nonhemolyzed serum should be used.

Certifications

FDA-cleared test (510k # K112422)

Specifications

Content And Storage 22°C to 25°C
Detectable Analytes Cryptococcal Antigen (CrAg)
Type Cryptococcus Antigen Assay Strips
Includes LF specimen diluent 2.5mL, CrAg LF test strips 50, CrAg positive control 1mL, package insert
Quantity 50 Tests
Test Time 10 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.