Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL201 Heavy Duty Anti-Vibration Mechanics Gloves with TPR Padding

Catalog No. 19555044 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-044 Small
19-555-043 Medium
19-555-042 Large
19-555-046 X-Large
19-555-045 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-044 Supplier Pyramex Safety Products Supplier No. GL201S
This item is not returnable. View return policy

Gloves feature TPR padding on three fingers for protection against impacts, and anti-shock EVA padding on the index finger.

  • Two-layer stretch spandex construction for comfort and flexibility
  • TPR padding on three fingers for impact protection
  • Anti-shock EVA padding on the index finger and palm
  • PVC patches on the palm for enhanced abrasion resistance
  • Soft terry cloth brow-wipe on the back of the glove
  • Hook and loop closure

Specifications

Back Material Spandex
Coating Material Uncoated
Color Black, Blue
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Small
EN388 Standard 2121
Finger Style Full
For Use With (Application) Carpentry, Construction, Machine Operation, and Mechanics
Glove Cuff Style Hook and Loop
Palm Type PVC Patches with EVA Padding
Series GL201
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.