Learn More
Ampco Safety Tools Impact Socket, 1/2 in. Drive (Imperial)
SDP
Catalog No. 19174004 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
1 1/2 in.
1 1/4 in.
1 1/8 in.
1 3/8 in.
1 5/8 in., 4 PT
1 7/16 in.
1 7/16 in., 4 PT
1 7/16 in., 8 PT
1 9/16 in.
11/16 in.
13/16 in.
15/16 in.
3/4 in.
3/8 in.
5/8 in.
7/16 in.
7/8 in.
9/16 in.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-174-004 3/8 in.
19-173-969 11/16 in.
19-173-971 1 1/2 in.
19-173-972 1 1/4 in.
19-173-973 1 1/8 in.
19-173-976 13/16 in.
19-173-978 1 3/8 in.
19-173-981 15/16 in.
19-173-983 1 5/8 in., 4 PT
19-173-986 1 7/16 in.
19-173-987 1 7/16 in., 4 PT
19-173-988 1 7/16 in., 8 PT
19-173-991 1 9/16 in.
19-174-003 3/4 in.
19-174-007 5/8 in.
19-174-008 7/16 in.
19-174-009 7/8 in.
19-174-011 9/16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-004 Supplier Ampco Safety Tools Supplier No. I1/2D3/8
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Impact Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Impact Socket Wrench
Drive Size 1/2 in.
Length (English) 3/8 in.
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.