Learn More
Ampco Safety Tools Impact Socket, 1/2 in. Drive (Metric)
SDP
Catalog No. 19173984 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
8 mm
9 mm
10 mm
11 mm
12 mm
13 mm
14 mm
15 mm
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
32 mm
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-173-984 15 mm
19-173-968 10 mm
19-173-974 11 mm
19-173-975 12 mm
19-173-979 13 mm
19-173-980 14 mm
19-173-985 16 mm
19-173-989 17 mm
19-173-990 18 mm
19-173-992 19 mm
19-173-993 20 mm
19-173-994 21 mm
19-173-995 22 mm
19-173-996 23 mm
19-173-997 24 mm
19-173-998 25 mm
19-173-999 26 mm
19-174-000 27 mm
19-174-001 28 mm
19-174-002 29 mm
19-174-005 30 mm
19-174-006 32 mm
19-174-010 8 mm
19-174-012 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-984 Supplier Ampco Safety Tools Supplier No. I1/2D15MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Impact Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Impact Socket Wrench
Drive Size 1/2 in.
Length (Metric) 15 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.