Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Three Quarter Finger Anti-Impact Nylon Gloves with VEP Padding

Catalog No. 19039260 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Hand:
Left
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hand
19-039260 Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-039260 Supplier Impacto™ Supplier No. 50510110041
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VEP padding with a suede leather cover on the palm provides protection from impact and abrasion. Nylon fabric on the back ensures breathability and comfort.

  • Nylon fabric ensures breathability and comfort
  • Contoured VEP padding in the palm provides protection from impact and shock
  • 3/4 finger design leaves finger tips open to for tactile feedback and mobility
  • Suede leather cover offers abrasion protection

Specifications

Back Material Nylon Lycra™
Color Blue, Gray
Hand Left
Material Suede
Product Type Anti-impact Gloves
Size Large
Finger Style Three Quarter
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Series 505-10
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.