Learn More
Impacto™ Air Temp Advantage Back Support
Impacto™ Air Temp Advantage with pockets supports the lower back helping to alleviate pain and discomfort
Supplier:  Impacto™ ATAXXXLRG
Catalog No. 19-031684
Only null left
Add to Cart
Description
Description
Impacto™ Air Temp Advantage Back Support
  • Two air chambers with self-contained aspirator provide support for the normal lordotic curve of the lower back
  • Belt has two pockets that rotate off center
  • Knit polyester elastic straps provide additional support
  • Neoprene on inside of belt enhances softness and flexibility
  • Breathable elastic across back
  • Non-metallic support stays with nonskid strips on outer stays
  • Fully adjustable closure
  • Specifications
    Specifications
    Hook and loop
    3X-Large
    Built in pockets, 2 inflatable air support chambers with self contained aspirator, Plastic support stays with non-slip strips
    Nylon
    144.78 to 157.48 cm
    Air Temp Advantage Belt
    Air chambers offer therapeutic pain relief, promotes muscle relaxation, improves posture alignment, helps alleviate back discomfort
    Neoprene inner
    57 to 62 in.
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.