Learn More
Impacto™ Air Belt Sport Plus Back Support
Impacto™ Air Belt Sports Plus back support offer natural therapy to support the back and help alleviate pain and discomfort
Supplier:  Impacto™ SPORTSPLUSXLARGE
Catalog No. 19-148-113
Only null left
Add to Cart
Description
Description
Impacto™ Air Belt Sports Plus
  • Breathable cotton outer cover
  • Unique patented inflatable air technology
  • Multi-celled air chambers conform to the back when air is added to provide relief from back pain.
  • Natural therapy and support
Specifications
Specifications
Hook and loop
X-Large
Inflatable multicelled air chamber, palm grip self contained aspirator with pressure release valve
Cotton
106.68 to 127 cm
Sports Plus Lumbar Support
Promotes muscle relaxation, improves posture alignment, naturally supports the lower back, helps alleviate pain and discomfort
Foam
42 to 50 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.