Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Vibration Nylon Gloves with Grain Leather Tool Grip

Catalog No. 19031966 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
Hand:
Left
Right
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-031966 Medium Right
19-031949 Large Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-031966 Supplier Impacto™ Supplier No. 50220110032
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VEP padding in the palm, thumb webbing, and index finger protects users from cumulative trauma caused by constant tool gripping.

  • Fingerless leather glove with stretchy nylon back
  • Soft grain leather palm cover for longer wear
  • Impact and shock absorbing VEP padding in the palm, between the thumb, and on the index finger
  • Thumb web area is protected from cumulative trauma such as bruising from gripping tools
  • Provides protection from impact, vibration, and abrasion
  • Ideal for use with hand tools and power tools

Specifications

Back Material Nylon
Color Blue, Yellow
Hand Right
Material Grain Leather
Product Type Tool Gloves
Size Medium
Finger Style Fingerless
For Use With (Application) Construction, DIY Home Improvement
Glove Cuff Style Open
Series 502-20
Length (English) 8 in.
Length (Metric) 20.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.