Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Three-Quarter Finger Anti-Impact Grain Leather Gloves

Catalog No. 19084728 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Hand:
Specific
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-084728 Small Specific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-084728 Supplier Impacto™ Supplier No. 50620110020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contoured VEP padding in the palm, fingers, and thumb provides protection from impact and shock. The open fingertip design ensures tactile feedback and mobility.

  • Grain leather on the front and back of the glove offers abrasion protection and durability
  • Gussetted fingers for additional comfort
  • Ideal for protection from hand, electric, and power tools
  • Use in applications such as construction, assembly, manufacturing, maintenance, and warehousing

Specifications

Color Blue, Yellow
Hand Specific
Material Grain Leather
Product Type Anti-impact Gloves
Size Small
Finger Style Three Quarter
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Series 506-20
Length (English) 7 in.
Length (Metric) 18 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.