Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Anti-Impact Trigger Finger Gloves with VEP Padding

Catalog No. 19048713D Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Hand:
Right
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-048-713D Medium Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-048-713D Supplier Impacto™ Supplier No. 40930 MD RH
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed of soft pearl leather to provide dexterity and abrasion resistance. Extended index finger design ensures tactile feedback and mobility.

  • Soft pearl leather palm provides dexterity and abrasion protection
  • Back is constructed of peal leather and nylon to ensure breathability
  • VEP 1/8 in. padding in the palm, thumb, and index finger for protection from impact and shock
  • Half finger design with extended index finger provides tactile feedback and mobility
  • Elastic wrist with hook and loop closure
  • Pull tabs at the fingertips for easy removal
  • Ideal for protection from electric, air, and power tools

Specifications

Cuff Material Elastic
Color Black, White
Hand Right
Material Pearl Leather
Product Type Anti-impact Gloves
Size Medium
Finger Style Half
For Use With (Application) Construction, Manufacturing and Maintenance
Glove Cuff Color Black
Glove Cuff Style Hook and Loop
Series 409-30
Length (English) 8 in.
Length (Metric) 20.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.