Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Cush'N Step Anti-Fatigue Flat Insoles

Catalog No. 19039352 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
5 to 6.5
7 to 8.5
Gender:
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Gender
19-039352 7 to 8.5 Women's
19-039349 5 to 6.5 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-039352 Supplier Impacto™ Supplier No. CUSHFLATB
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Impacto™ Cush'N Step CUSHFLAT Anti-fatigue insoles provide full foot comfort and cushioning with low cut and dress shoes

Impacto™ Cush'N Step Anti-Fatigue Insoles

  • Cellular cushioning open cell foam
  • Biocide additive helps prevent foot odor
  • Full flat design for use with dress and low cut footwear
  • Helps relieve foot, legs and back fatigue
  • Great for daily wear with low cut and dress shoes

Specifications

Color Green
For Use With (Application) Use with dress and low cut footwear
Length (English) 8 in.
Length (Metric) 20.32 cm
Material Open-cell Foam
Size 7 to 8.5
Style Flat
Gender Women's
Product Type Full-tapered Insole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.