Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Cush'N Step Anti-Fatigue Molded Insoles

Catalog No. 19062962 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
5 to 6.5
7 to 8.5
Gender:
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Gender
19-062962 7 to 8.5 Women's
19-062960 5 to 6.5 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-062962 Supplier Impacto™ Supplier No. CUSHMOLDB
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Impacto™ Cush'N Step CUSHMOLD is Made of lightweight cellular cushioning open-cell foam with re-reinforced gel inserts in the heel

Impacto™ Cush'N Step Anti-Fatigue Insoles

  • Cellular cushioning open cell foam with antibacterial fabric cover
  • Molded design with GEL heel insert and arch support
  • Helps relieve foot, legs and back fatigue
  • Pain and fatigue relief when standing or walking on hard, cold surfaces for long periods
  • For use with closed shoes, boots and sports shoes

† Also available in case of 12 packs.

Specifications

Color Blue
For Use With (Application) Sports shoes, work boots and work footwear
Length (English) 8 in.
Length (Metric) 20.32 cm
Material Polymeric Open-cell Foam
Size 7 to 8.5
Style Molded
Gender Women's
Product Type Molded Insole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.