Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Double Padded Anti-Impact Gloves

Catalog No. 189994606D Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
Hand:
Left
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
18-999-4606D X-Large Left
18-999-4606C Large Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-4606D Supplier Impacto™ Supplier No. 52324110051
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nylon material offers breathability and comfort, while suede leather provides abrasion protection and durability. 1/4 in. VEP padding in the palm extends to the edge of the hand for protection from impact and shock.

  • VEP provides impact absorbing and shock dissipation qualities, as well as absorption of higher levels of vibration
  • Fingerless style for tactile feedback and mobility
  • Suede leather offers abrasion protection & durability
  • Side padding protects from impact during automotive applications

Specifications

Color Blue, Yellow
Hand Left
Material Nylon Lycra
Product Type Anti-impact Gloves
Size X-Large
Finger Style Fingerless
For Use With (Application) Automobile Construction
Glove Cuff Style Open
Series 523-24
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.