Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Nylon Gloves with Grain Leather Tool Grip

Catalog No. 19048715D Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-715D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-048-715D Supplier Impacto™ Supplier No. 50220PXL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contoured VEP padding in the palm and thumb webbing provides protection from impact and shock. The open fingertip design ensures tactile feedback and mobility.

  • Soft grain leather palm and thumb webbing with VEP padding
  • Thumb web area is protected from cumulative trauma such as bruising from gripping tools
  • Provides protection from impact, vibration, and abrasion
  • Use with hand tools and power tools

Specifications

Back Material Nylon
Color Blue, Yellow
Hand Specific
Material Grain Leather
Product Type Tool Gloves
Size X-Large
Finger Style Fingerless
For Use With (Application) Hand Tools and Power Tools
Glove Cuff Style Open
Series 502-20
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.