Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Anti-Impact Glove liners with wrist support

Catalog No. 19039276 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-039276 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039276 Supplier Impacto™ Supplier No. 71420120030
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Impacto™ 714-20 Glove liner with padding in the palm and thumb web protects from impact and vibration and wrist support protects from RSI

Impacto™ Anti-Impact Glove Liner With Wrist Support

  • Fingerless, polycotton
  • VEP padding in the palm and thumb web area
  • Detachable grain leather wrist support minimizes extension and over flexion of the wrist to prevent RSI such as carpal tunnel syndrome or tendinitis
  • Provides protection from abrasion, impact and vibration
  • Fingerless style for optimum dexterity
  • Used with hand or electric tools in repetitive tasks

Specifications

Color Blue, Gold
Closure Type Hook and loop
Product Type Fingerless Glove Liner
Design Fingerless
Length (Metric) 22.86 cm
Length (English) 9 in.
Size Medium
For Use With (Application) Provides protection from impact and shock and recommended to be worn inside specialty glove for maximum durability
Includes Detachable grain Leather wrist support, VEP palm padding
Hand Specific
Material Polycotton
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.