Learn More
Impacto™ Anti-Impact Glove liners with wrist support
Impacto™ 714-20 Glove liner with padding in the palm and thumb web protects from impact and vibration and wrist support protects from RSI
Supplier:  Impacto™ 71420120050
 View more versions of this product
Catalog No. 19-039281
Only null left
Add to Cart
Description
Description
Impacto™ Anti-Impact Glove Liner With Wrist Support
  • Fingerless, polycotton
  • VEP padding in the palm and thumb web area
  • Detachable grain leather wrist support minimizes extension and over flexion of the wrist to prevent RSI such as carpal tunnel syndrome or tendinitis
  • Provides protection from abrasion, impact and vibration
  • Fingerless style for optimum dexterity
  • Used with hand or electric tools in repetitive tasks
Specifications
Specifications
Blue, Gold
Fingerless Glove Liner
22.86 cm
X-Large
Detachable grain Leather wrist support, VEP palm padding
Polycotton
Hook and loop
Fingerless
9 in.
Provides protection from impact and shock and recommended to be worn inside specialty glove for maximum durability
Specific
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.