Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Anti-Impact Glove Liners with Extended VEP Padding

Catalog No. 189994602A Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Hand:
Left
Right
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
18-999-4602A Small Left
18-999-4601A Small Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-4602A Supplier Impacto™ Supplier No. 51000120021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stretch polycotton fabric ensures optimum breathability and comfort. VEP padding that extends from the palm to the wrist joint provides users with additional protection.

  • 4-way stretch polycotton fabric ensures optimum breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm extends to the wrist joint for additional protection
  • Open-finger design for tactile feedback and mobility
  • Designed to be worn under work gloves
  • Offers protection from electric, manual, and power tools
  • Use with applications such as construction, assembly, manufacturing, maintenance, and warehousing

Specifications

Color Blue
Hand Left
Material Polycotton
Product Type Glove Liners
Size Small
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Series 510-00
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.