Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Nylon Gloves with Detachable Wrist Support

Catalog No. 19031838 Shop All Impacto Protective Products Products
Encompass_Preferred
Change view
Click to view available options
Cuff Material:
Grain Leather
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cuff Material
19-031838 Grain Leather
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-031838 Supplier Impacto™ Supplier No. 70320120040
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grain leather palm offers durability and abrasion resistance, while detachable wrist support protects from impact and repetitive strain injuries.

  • Stretch Nylon Lycra™ material offers breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm protects from impact
  • Fingerless style for tactile feedback and mobility
  • Grain leather offers abrasion protection and durability
  • Detachable leather wrist support prevents over-flexion and over extension to prevent repetitive strain injuries

Specifications

Cuff Material Grain Leather
Color Blue, Yellow
Hand Specific
Material Nylon Lycra
Product Type Anti-impact Gloves
Size Large
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, and Maintenance
Glove Cuff Color Yellow
Series 703-20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.