Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Gloves with Detachable Leather Wrist Support

Catalog No. 19031799 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
Hand:
Specific
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-031799 X-Large Specific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-031799 Supplier Impacto™ Supplier No. 70310120050
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nylon fabric ensures that gloves are breathable and comfortable. Detachable suede leather wrist support helps to prevent over flexion and over extension.

  • Nylon fabric ensures optimum breathability and comfort
  • Contoured VEP 1/8 in. padding in palm and wrist
  • Fingerless style leaves fingers open for tactile feedback and mobility
  • Detachable wrist support helps to prevent repetitive strain injuries
  • Offers protection from hand tools and power tools
  • Ideal for applications in construction, assembly, manufacturing, maintenance, and warehousing

Specifications

Cuff Material Suede Leather
Color Blue, Gray
Hand Specific
Material Nylon
Product Type Anti-impact Gloves
Size X-Large
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Color Gray
Series 703-10
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.